Få mere at vide
11alpha-Hydroxy Progesterone, TRC
Artikelnummer. 30136202
Skift visning
Mængde:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136202 1 g
30133613 5 g
17942515 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136202 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9523351G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 80-75-1
Molekylær formel C21 H30 O3
Molekylvægt (g/mol) 330.46
Synonym 11alpha-Hydroxyprogesterone, Pregn-4-ene-3,20-dione, 11-hydroxy-, (11α)- (9CI, ACI), (11α)-11-Hydroxypregn-4-ene-3,20-dione (ACI), Pregn-4-ene-3,20-dione, 11α-hydroxy- (8CI), Progesterone, 11α-hydroxy- (5CI), 11-Hydroxyprogesterone, 11α-Hydroxypregn-4-ene-3,20-dione, 11α-Hydroxyprogesterone, 4-Pregnene-11α-ol-3,20-dione, DG 322B, NSC 3350, Pregn-4-en-11α-ol-3,20-dione, U 0384
IUPAC navn (8S,9S,10R,11R,13S,14S,17S)-17-acetyl-11-hydroxy-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
SMIL CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@H](O)C[C@]12C
Tekniske data
Kemisk navn eller materiale 11a-Hydroxyprogesterone
Mængde 1 g
InChI formel InChI=1S/C21H30O3/c1-12(22)16-6-7-17-15-5-4-13-10-14(23)8-9-20(13,2)19(15)18(24)11-21(16,17)3/h10,15-19,24H,4-9,11H2,1-3H3/t15-,16+,17-,18+,19+,20-,21+/m0/s1
Formel vægt 330.2195