Få mere at vide
11-O-Tetrahydropyranyl-15-O-tert-butyldimethylsilyl-lubiprostone-d7 Phenylmethyl Ester, TRC
Artikelnummer. 30420584
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30420584 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420584 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2950521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 11-O-Tetrahydropyranyl-15-O-tert-butyldimethylsilyl-lubiprostone-d7 Phenylmethyl Ester
CAS 1346597-45-2
Molekylær formel C38H55D7F2O6Si
Mængde 1 mg
Synonym (11alpha)-16,16-Difluoro-11-O-tetrahydropyranyl-15-O-tert-butyldimethylsilyl-9-oxoprostan Phenylmethyl Ester-d7
InChI formel InChI=1S/C38H62F2O6Si/c1-7-8-25-38(39,40)34(46-47(5,6)37(2,3)4)24-23-31-30(32(41)27-33(31)45-36-22-16-17-26-43-36)20-14-9-10-15-21-35(42)44-28-29-18-12-11-13-19-29/h11-13,18-19,30-31,33-34,36H,7-10,14-17,20-28H2,1-6H3/t30-,31-,33-,34?,36?/m1/s1/i1D3,7D2,8D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])CC(F)(F)C(CC[C@H]1[C@@H](CC(=O)[C@@H]1CCCCCCC(=O)OCc2ccccc2)OC3CCCCO3)O[Si](C)(C)C(C)(C)C
IUPAC navn benzyl 7-[(1R,2R,3R)-2-[3-[tert-butyl(dimethyl)silyl]oxy-6,6,7,7,8,8,8-heptadeuterio-4,4-difluorooctyl]-3-(oxan-2-yloxy)-5-oxocyclopentyl]heptanoate
Molekylvægt (g/mol) 688.02
Formel vægt 687.4723