Få mere at vide
11-Ketoprogesterone, Mikromol™
Artikelnummer. 17957714
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17957714 100 mg
17927634 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17957714 Leverandør Mikromol Leverandørnr. MM0222.12
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
11-Ketoprogesterone
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 11-Ketoprogesterone
CAS 516-15-4
Analyt- eller komponentnavne 11-Ketoprogesterone
Forsendelsestilstand Room Temperature
Molekylær formel C21 H28 O3
Certificeringer/compliance ISO 9001
Synonym 11-Ketoprogesterone
InChI formel InChI=1S/C21H28O3/c1-12(22)16-6-7-17-15-5-4-13-10-14(23)8-9-20(13,2)19(15)18(24)11-21(16,17)3/h10,15-17,19H,4-9,11H2,1-3H3/t15-,16+,17-,19+,20-,21+/m0/s1
SMIL CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C
IUPAC navn (8S,9S,10R,13S,14S,17S)-17-acetyl-10,13-dimethyl-2,6,7,8,9,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthrene-3,11-dione
Molekylvægt (g/mol) 328.45
Mængde 100 mg
Formel vægt 328.2038 g/mol