Få mere at vide
11-Deoxy Corticosterone Pivalate, TRC
Artikelnummer. 30359360
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359360 100 mg
30359644 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30359360 Leverandør Toronto Research Chemicals Leverandørnr. TRCD232580100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 808-48-0
Molekylær formel C26 H38 O4
Molekylvægt (g/mol) 414.58
Synonym 21-(2,2-Dimethyl-1-oxopropoxy)pregn-4-ene-3,20-dione, 11-Deoxycorticosterone pivalate, Deoxycorticosterone pivalate, 11-Deoxycorticosterone Pivalic acid ester with, 21-Hydroxypregn-4-ene-3,20-dione pivalate, Cortexone M, DOCP, DTMA, Deoxycorticosterone pivalate, Deoxycorticosterone trimethylacetate, Deoxycortone pivalate, Deoxycortone trimethylacetate, Desoxycorticosterone pivalate, Desoxycorticosterone trimethylacetate, Desoxycortisone pivalate, Desoxycortone pivalate, NSC 95278, Neodin-Depositum, Percorten M, Percorten Pivalate
IUPAC navn [2-[(8S,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2-dimethylpropanoate
SMIL CC(C)(C)C(=O)OCC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale Desoxycorticosterone Pivalate
Mængde 100 mg
InChI formel InChI=1S/C26H38O4/c1-24(2,3)23(29)30-15-22(28)21-9-8-19-18-7-6-16-14-17(27)10-12-25(16,4)20(18)11-13-26(19,21)5/h14,18-21H,6-13,15H2,1-5H3/t18-,19-,20-,21+,25-,26-/m0/s1
Formel vægt 414.277