Få mere at vide
11-Dehydrodexamethasone, TRC
Artikelnummer. 30134870
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134870 50 mg
30007628 100 mg
30006698 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134870 Leverandør Toronto Research Chemicals Leverandørnr. TRCD35736550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2964-81-0
Molekylær formel C22 H27 F O5
Molekylvægt (g/mol) 390.4452
Synonym Pregna-1,4-diene-3,11,20-trione, 9-fluoro-17,21-dihydroxy-16-methyl-, (16α)-, Pregna-1,4-diene-3,11,20-trione, 9-fluoro-17,21-dihydroxy-16α-methyl- (7CI,8CI), (16α)-9-Fluoro-17,21-dihydroxy-16-methylpregna-1,4-diene-3,11,20-trione, 11-Dehydrodexamethasone, 9-Fluoro-17,21-dihydroxy-16α-methylpregna-1,4-diene-3,11,20-trione
IUPAC navn (8S,9R,10S,13S,14S,16R,17R)-9-fluoro-17-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthrene-3,11-dione
SMIL C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)C(=O)C[C@]2(C)[C@@]1(O)C(=O)CO
Tekniske data
Kemisk navn eller materiale 11-Dehydrodexamethasone
Mængde 50 mg
InChI formel InChI=1S/C22H27FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-16,24,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,19+,20+,21+,22+/m1/s1
Formel vægt 390.1843