missing translation for 'onlineSavingsMsg'
Få mere at vide

11-Dehydro-6Alpha,17-dihydroxy-corticosterone 21-Acetate, TRC

Artikelnummer. 30367599
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367599 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30367599 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23031350MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 11-Dehydro-6alpha,17-dihydroxy-corticosterone 21-Acetate
CAS 66978-87-8
Molekylær formel C23H30O7
Mængde 50 mg
Synonym (6alpha)-21-(Acetyloxy)-6,17-dihydroxy-pregn-4-ene-3,11,20-trione, 2-((6S,8S,9S,10R,13S,14S,17R)-6,17-Dihydroxy-10,13-dimethyl-3,11-dioxo-2,3,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)-2-oxoethyl Acetate
InChI formel InChI=1S/C23H30O7/c1-12(24)30-11-19(28)23(29)7-5-15-14-9-17(26)16-8-13(25)4-6-21(16,2)20(14)18(27)10-22(15,23)3/h8,14-15,17,20,26,29H,4-7,9-11H2,1-3H3/t14-,15-,17-,20+,21-,22-,23-/m0/s1
SMIL CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3C[C@H](O)C4=CC(=O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C
IUPAC navn [2-[(6S,8S,9S,10R,13S,14S,17R)-6,17-dihydroxy-10,13-dimethyl-3,11-dioxo-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molekylvægt (g/mol) 418.48
Formel vægt 418.1992
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.