missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ 10X TdT Labeling Buffer

Artikelnummer. 30000909
Klik for at se tilgængelige muligheder
Mængde:
20 ml
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 30000909

Brand: R&D Systems™ 48176002

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

R&D Systems™ 10X TdT Labeling Buffer is a concentrated and safe, cacodylate-free solution developed for use in all TUNEL-based applications.

  • Long Name: Terminal Deoxynucleotidyl Transferase
  • Do not use past expiration date
  • There are no hazardous ingredients present at or above the reporting threshold as defined in the United States 29 CFR 1910.1200 (OSHA) or the Canadian Controlled Products Regulations (WHMIS).
  • Store in a dry, cool, and well-ventilated place
  • Keep container closed when not in use
  • Keep/Store away from direct sunlight, extremely high or low temperatures, and incompatible materials
  • Ingestion is likely to be harmful or have adverse effects
  • Uncontrolled product according to WHMIS classification criteria
TRUSTED_SUSTAINABILITY

Specifications

Mængde 20 ml
Indhold og opbevaring Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.