Få mere at vide
Medchem Express 10mM * 1 mL PF-CBP1 hydrochloride
Brand:  Medchem Express HY-19999A_1mL
Encompass_Preferred
10mM * 1 mL PF-CBP1 hydrochloride
Produktkode 18620303
  • Ring for pris / 1ml
 Begrænset vare
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Se alternative produkter »