Learn More
Medchem Express 10mM * 1 mL PF-03463275
Brand:  Medchem Express HY-10716A_1mL
Encompass_Preferred
10mM * 1 mL PF-03463275
Product Code. 18612113
  • 1714.26 DKK / 1ml
 Restricted Item
Forventet forsendelse: 24-11-2026