Learn More
Chemsplash 10gsm Mop Cap Pleated Double Stitched
The Double Stitched Disposable Mob Cap is made from a 10GSM blue non-woven polypropylene breathable material, which is elasticated at the rim for ease of fit to your head.
595.00 DKK
Specifications
Product Type | Mob Cap |
---|---|
Size | 21 in. |
Material | Polypropylene |
Disposable | Single-use |
Latex Free | Yes |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
30058153
![]() |
Chemsplash
2516.300.999 |
Blue |
595.00 DKK
Pack of 2000 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
30058162
![]() |
Chemsplash
2516.400.999 |
White |
595.00 DKK
Pack of 2000 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
30058226
![]() |
Chemsplash
2516.500.999 |
Red |
595.00 DKK
Pack of 2000 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
30058270
![]() |
Chemsplash
2516.600.999 |
Green |
595.00 DKK
Pack of 2000 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
30058149
![]() |
Chemsplash
2516.700.999 |
Yellow |
595.00 DKK
Pack of 2000 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
Description
Double Stitched Disposable Mob Cap
- 10 Gram Non Woven Polypropylene
- Double Stitched
- Elasticated
- 21 in.
- One Size
- PFAS Free
Description: The Double Stitched Disposable Mob Cap is made from a 10GSM blue non-woven polypropylene breathable material, which is elasticated at the rim for ease of fit to your head. Double-stitched for extra strength, and 21 in. in size. Suitable for all industries requiring disposable head covering.
Specifications
Mob Cap | |
Polypropylene | |
Yes |
21 in. | |
Single-use | |
Non-sterile |