Få mere at vide
(10E)-10-Octadecenoic acid, TRC
Artikelnummer. 30179390
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30179390 25 mg -
30179111 2.5 mg 2.5mg
30179005 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179390 Leverandør Toronto Research Chemicals Leverandørnr. TRCO23525025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5684-82-2
Molekylær formel C18H34O2
Molekylvægt (g/mol) 282.46
Synonym Isooleic Acid
IUPAC navn (E)-octadec-10-enoic acid
SMIL CCCCCCC\C=C\CCCCCCCCC(=O)O
Tekniske data
Kemisk navn eller materiale (10E)-10-Octadecenoic acid
Mængde 25 mg
InChI formel InChI=1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h8-9H,2-7,10-17H2,1H3,(H,19,20)/b9-8+
Formel vægt 282.2559