Få mere at vide
10Alpha-Methoxy-1-methyl-9,10-dihydrolysergol, TRC
Artikelnummer. 30469460
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469460 2.5 mg
30475203 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30469460 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2622952.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 35155-28-3
Molekylær formel C18 H24 N2 O2
Molekylvægt (g/mol) 300.4
Synonym [(6aR,9R,10aS)-10a-Methoxy-4,7-dimethyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinolin-9-yl]methanol, Nicergoline Imp. C (EP), Methylmethoxylumilysergol, Ergoline-8-methanol, 10-methoxy-1,6-dimethyl-, (8β)-, Lumilysergol, 10O,1-dimethyl- (7CI), (8β)-10-Methoxy-1,6-dimethylergoline-8-methanol, Indolo[4,3-fg]quinoline, ergoline-8-methanol deriv., (+)-(5R,10S)-Meluol, 1,6-Dimethyl-8β-(hydroxymethyl)-10α-methoxyergoline, 1-Methyl-10α-methoxydihydrolysergol, 10-Methoxy-1,6-dimethylergoline-8β-methanol, 10α-Methoxy-1-methyl-9,10-dihydrolysergol, 10α-Methoxy-1-methyldihydrolysergol, O10,1-Dimethyllumilysergol
IUPAC navn [(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methanol
SMIL CO[C@]12C[C@@H](CO)CN(C)[C@@H]1Cc3cn(C)c4cccc2c34
Tekniske data
Kemisk navn eller materiale Methylmethoxylumilysergol
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C18H24N2O2/c1-19-10-13-7-16-18(22-3,8-12(11-21)9-20(16)2)14-5-4-6-15(19)17(13)14/h4-6,10,12,16,21H,7-9,11H2,1-3H3/t12-,16-,18+/m1/s1
Formel vægt 300.1838
Procent renhed >95%