Få mere at vide
Paragon Scientific 100ML Sulfur @ 0.0025% (25ppm) in Isoo..ctane
Brand:  Paragon Scientific ALK-IOSU-030
100ML Sulfur @ 0.0025% (25ppm) in Isoo..ctane
Produktkode 30457500
  • Ring for pris / 100ml
 Begrænset vare
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Se alternative produkter »