Learn More
Reagecon 100ML ETALON DENSITE 0.7488G/ML @20°C

Brand: Reagecon DEN20060
447.61 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
100ML ETALON DENSITE 0.7488G/ML @20°C
Safety and Handling
- DENSITY STANDARD 0.7488g/ml @ 20°C
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : dodecane
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
DEN20624G1 | Certificate of Analysis | 01/12/2024 | DEN20060 |
DEN20623A1 | Certificate of Analysis | 23/07/2024 | DEN20060 |
DEN20623M1 | Certificate of Analysis | 08/05/2024 | DEN20060 |
DEN20622B1 | Certificate of Analysis | 17/05/2022 | DEN20060 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.