Learn More
10-Undecyn-1-ol, 96%, Thermo Scientific Chemicals
Description
10-Undecyn-1-ol exhibits an antifungal activity. It is also used in the esterification of pentanoic and stearic acids in the presence of lipase. It is also employed as a reagent in the preparation of 11-bromo-undec-1-yne by reacting with carbon tetrabromide and triphenylphosphine. Further, it acts as an intermediate in synthetic chemistry. In addition to this, it serves as a precursor in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
10-Undecyn-1-ol exhibits an antifungal activity. It is also used in the esterification of pentanoic and stearic acids in the presence of lipase. It is also employed as a reagent in the preparation of 11-bromo-undec-1-yne by reacting with carbon tetrabromide and triphenylphosphine. Further, it acts as an intermediate in synthetic chemistry. In addition to this, it serves as a precursor in organic synthesis.
Solubility
Immiscible with water.
Notes
Air sensitive. Incompatible strong oxidizing agents and strong bases.
Specifications
Density | 0.873 |
Boiling Point | 110°C to 111°C (4 mmHg) |
Refractive Index | 1.4565 |
Quantity | 1 g |
Beilstein | 1702764 |
Solubility Information | Immiscible with water. |
Formula Weight | 168.28 |
Percent Purity | 96% |
Chemical Name or Material | 10-Undecyn-1-ol |
RUO – Research Use Only