missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
10-trans,12-cis-Linoleic Acid, TRC
Specifications
Specifications
| Kemisk navn eller materiale | 10-trans,12-cis-Linoleic Acid |
| CAS | 2420-56-6 |
| Molekylær formel | C18 H32 O2 |
| Mængde | 500 mg |
| Synonym | 10,12-Octadecadienoic acid, (Z,E)-, (10E,12Z)-10,12-Octadecadienoic acid, 10-trans,12-cis-Linoleic acid, 10-trans-12-cis-Octadecadienoic acid, 10E,12Z-Octadecadienoic acid, 10t12c Conjugated linoleic acid, C18:2 t-10,c-12, trans-10,cis-12 Conjugated linoleic acid, trans-10-cis-12-Octadecadienoic acid, 10,12-Octadecadienoic acid, (10E,12Z)-, (10E,12Z)-10,12-Octadecadienoic Acid |
| InChI formel | InChI=1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-9H,2-5,10-17H2,1H3,(H,19,20)/b7-6-,9-8+ |
| SMIL | CCCCC\C=C/C=C/CCCCCCCCC(=O)O |
| IUPAC navn | (10E,12Z)-octadeca-10,12-dienoic acid |
| Molekylvægt (g/mol) | 280.45 |
| Formel vægt | 280.2402 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?