missing translation for 'onlineSavingsMsg'
Få mere at vide

10-(Pentafluorophenoxycarbonyl)decyltriethoxysilane, 95%

Artikelnummer. 15428755
Change view
Click to view available options
Mængde:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
15428755 250 mg 250mg
15418755 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428755 Supplier Thermo Scientific Alfa Aesar Supplier No. H34037.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1197981-08-0 | C23H35F5O5Si | 514.605 g/mol

Denne Thermo Scientific Chemicals mærkevare var oprindeligt en del af Alfa Aesar produktportefølje. Noget dokumentation og etiketoplysninger kan referere til det gamle mærke. Originalen Alfa Aesar produkt-/varekode eller SKU-reference er ikke ændret som en del af mærkeovergangen til Thermo Scientific Chemicals .

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197981-08-0
Molekylær formel C23H35F5O5Si
Molekylvægt (g/mol) 514.605
MDL nummer MFCD13181916
InChI nøgle WQFFEEIHQJVCHR-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorophenyl 11-triethoxysilyl undecanoate, 10-pentafluorophenoxycarbonyl decyltriethoxysilane, 10-pentafluorophenoxycarbonyl decyl triethoxysilane
PubChem CID 99738205
IUPAC navn (2,3,4,5,6-pentafluorphenyl) 11-triethoxysilylundecanoat
SMIL CCO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC

Specifications

Kogepunkt 155°C to 160°C (0.04mmHg)
Kemisk navn eller materiale 10-(Pentafluorophenoxycarbonyl)decyltriethoxysilane
Mængde 250 mg
Formel vægt 514.61
Procent renhed 95%
Brydningsindeks 1.4320-1.4360
Følsomhed Moisture sensitive

RUO– Kun brug til forskning

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.