missing translation for 'onlineSavingsMsg'
Få mere at vide

10-Methylundecanol, TRC

Artikelnummer. 30480532
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30480532 2.5 g 2.5g
30469920 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Product Code. 30480532 Supplier Toronto Research Chemicals Supplier No. TRCM3381102.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20194-45-0
Molekylær formel C12H26O
Molekylvægt (g/mol) 186.33
Synonym 10-Methyl-1-undecanol
SMIL CC(C)CCCCCCCCCO

Specifications

Kemisk navn eller materiale 10-Methylundecanol
Mængde 2.5 g
InChI formel InChI=1S/C12H26O/c1-12(2)10-8-6-4-3-5-7-9-11-13/h12-13H,3-11H2,1-2H3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.