missing translation for 'onlineSavingsMsg'
Learn More

10-Methylphenothiazine, 98%, Thermo Scientific Chemicals

Catalog Number p-7025916
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1207-72-3
C13H11NS
213.30
MFCD00041836
QXBUYALKJGBACG-UHFFFAOYSA-N
10-methyl-10h-phenothiazine, n-methylphenothiazine, 10h-phenothiazine, 10-methyl, phenothiazine, 10-methyl, acmc-1bo1o, bidd:gt0308, phenothiazide methyl derivative, 10-methylphenothiazine, 10-methyl-10h-phenothiazine #, phenothiazine, 10-methyl-8ci
71015
10-methylphenothiazine
CN1C2=CC=CC=C2SC2=CC=CC=C12
This item is not returnable. View return policy
1207-72-3
C13H11NS
213.30
MFCD00041836
QXBUYALKJGBACG-UHFFFAOYSA-N
10-methyl-10h-phenothiazine, n-methylphenothiazine, 10h-phenothiazine, 10-methyl, phenothiazine, 10-methyl, acmc-1bo1o, bidd:gt0308, phenothiazide methyl derivative, 10-methylphenothiazine, 10-methyl-10h-phenothiazine #, phenothiazine, 10-methyl-8ci
71015
10-methylphenothiazine
CN1C2=CC=CC=C2SC2=CC=CC=C12

They are used in pharmaceutical manufacturing especially antipsychotic drugs. Also used as a stabilizer against polymerization of acrylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are used in pharmaceutical manufacturing especially antipsychotic drugs. Also used as a stabilizer against polymerization of acrylates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 100°C to 103°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 213.3
Percent Purity 98%
Chemical Name or Material 10-Methylphenothiazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'