Få mere at vide
10-Methoxy-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-one, Mikromol™
Artikelnummer. 30050483
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050483 100 mg
30049941 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050483 Leverandør Mikromol Leverandørnr. MM0180.11
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
10-Methoxy-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 10-Methoxy-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-one
CAS 59743-84-9
Analyt- eller komponentnavne 10-Methoxy-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-one
Forsendelsestilstand Room Temperature
Molekylær formel C14 H10 O2 S
Certificeringer/compliance ISO 9001
Synonym 10-Methoxy-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-one
InChI formel InChI=1S/C14H10O2S/c1-16-12-8-9-4-2-3-5-10(9)13(15)11-6-7-17-14(11)12/h2-8H,1H3
SMIL COC1=Cc2ccccc2C(=O)c3ccsc13
IUPAC navn 4-methoxybenzo[1,2]cyclohepta[2,4-b]thiophen-10-one
Molekylvægt (g/mol) 242.29
Mængde 100 mg
Formel vægt 242.0402 g/mol