Få mere at vide
10-Hydroxyiminodibenzyl, Mikromol™
Artikelnummer. 30052081
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052081 25 mg
30051403 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052081 Leverandør Mikromol Leverandørnr. MM0076.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
10-Hydroxyiminodibenzyl
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 10-Hydroxyiminodibenzyl
CAS 4014-77-1
Analyt- eller komponentnavne 10-Hydroxyiminodibenzyl
Forsendelsestilstand Room Temperature
Molekylær formel C14 H13 N O
Certificeringer/compliance ISO 17025
Synonym 10,11-Dihydro-5H-dibenz[b,f]azepin-10-ol, 10-Hydroxyiminodibenzyl
InChI formel InChI=1S/C14H13NO/c16-14-9-10-5-1-3-7-12(10)15-13-8-4-2-6-11(13)14/h1-8,14-16H,9H2
SMIL OC1Cc2ccccc2Nc3ccccc13
IUPAC navn 6,11-dihydro-5H-benzo[b][1]benzazepin-5-ol
Molekylvægt (g/mol) 211.26
Mængde 25 mg
Formel vægt 211.0997 g/mol