Få mere at vide
10-Hydroxy Warfarin(Mixture of Diastereomers), TRC
Artikelnummer. 30134943
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134943 50 mg
30008075 5 mg
30009562 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134943 Leverandør Toronto Research Chemicals Leverandørnr. TRCH99616050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 83219-99-2
Molekylær formel C19 H16 O5
Molekylvægt (g/mol) 324.33
Synonym 2H-1-Benzopyran-2-one, 4-hydroxy-3-(2-hydroxy-3-oxo-1-phenylbutyl)-, 4-Hydroxy-3-(2-hydroxy-3-oxo-1-phenylbutyl)-2H-1-benzopyran-2-one, 10-Hydroxy warfarin, 10-Hydroxywarfarin
IUPAC navn 4-hydroxy-3-(2-hydroxy-3-oxo-1-phenylbutyl)chromen-2-one
SMIL CC(=O)C(O)C(C1=C(O)c2ccccc2OC1=O)c3ccccc3
Tekniske data
Kemisk navn eller materiale 10-Hydroxy Warfarin (Mixture of Diastereomers)
Mængde 50 mg
InChI formel InChI=1S/C19H16O5/c1-11(20)17(21)15(12-7-3-2-4-8-12)16-18(22)13-9-5-6-10-14(13)24-19(16)23/h2-10,15,17,21-22H,1H3
Formel vægt 324.0998