Få mere at vide
10-Heneicosanone, TRC
Artikelnummer. 30138228
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138228 50 mg
30135533 10 mg
17939834 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138228 Leverandør Toronto Research Chemicals Leverandørnr. TRCH10539050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22026-16-0
Molekylær formel C21H42O
Molekylvægt (g/mol) 310.56
IUPAC navn henicosan-10-one
SMIL CCCCCCCCCCCC(=O)CCCCCCCCC
Tekniske data
Kemisk navn eller materiale 10-Heneicosanone
Mængde 50 mg
InChI formel InChI=1S/C21H42O/c1-3-5-7-9-11-12-14-16-18-20-21(22)19-17-15-13-10-8-6-4-2/h3-20H2,1-2H3
Formel vægt 310.3236