Få mere at vide
10-Deacetyl-7-xylosyl Paclitaxel (62%), TRC
Artikelnummer. 30349969
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30349969 50 mg
30362044 10 mg
30351520 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30349969 Leverandør Toronto Research Chemicals Leverandørnr. TRCD19896550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 90332-63-1
Molekylær formel C50 H57 N O17
Molekylvægt (g/mol) 943.98
Synonym Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-6,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-4-(β-D-xylopyranosyloxy)-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, 12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-6,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-4-(β-D-xylopyranosyloxy)-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aα,4β,4aβ,6β,9α(αR*,βS*),11α,12α,12aα,12bα]]-, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete, benzenepropanoic acid deriv., 10-Deacetyl-7-xylosylpaclitaxel, 10-Deacetyl-7-xylosyltaxol, 10-Deacetylpaclitaxel 7-xyloside, 10-Deacetyltaxol 7-xyloside, 7-Xylosyl-10-deacetylpaclitaxel, 7-Xylosyl-10-deacetyltaxol, 7β-Xylosyl-10-deacetyltaxol
IUPAC navn [(1S,2S,3R,4S,7R,9S,10S,12R,15S)-4-acetyloxy-15-[(2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoyl]oxy-1,12-dihydroxy-10,14,17,17-tetramethyl-11-oxo-9-[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] benzoate
SMIL CC(=O)O[C@@]12CO[C@@H]1C[C@H](O[C@@H]3OC[C@@H](O)[C@H](O)[C@H]3O)[C@]4(C)[C@@H]2[C@H](OC(=O)c5ccccc5)[C@]6(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)c7ccccc7)c8ccccc8)C(=C([C@@H](O)C4=O)C6(C)C)C
Tekniske data
Kemisk navn eller materiale 10-Deacetyl-7-xylotaxol
Mængde 50 mg
InChI formel InChI=1S/C50H57NO17/c1-25-31(65-45(61)38(56)35(27-15-9-6-10-16-27)51-43(59)28-17-11-7-12-18-28)22-50(62)42(67-44(60)29-19-13-8-14-20-29)40-48(5,41(58)37(55)34(25)47(50,3)4)32(21-33-49(40,24-64-33)68-26(2)52)66-46-39(57)36(54)30(53)23-63-46/h6-20,30-33,35-40,42,46,53-57,62H,21-24H2,1-5H3,(H,51,59)/t30-,31+,32+,33-,35+,36+,37-,38-,39-,40+,42+,46+,48-,49+,50-/m1/s1
Formel vægt 943.3626