missing translation for 'onlineSavingsMsg'
Få mere at vide

10,12-tricosadiynosyre, TRC

Product Code. 30455535
Change view
Click to view available options
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30455535 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455535 Supplier Toronto Research Chemicals Supplier No. TRCT8964601MG
 Begrænset vare
Forventet forsendelse: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 10,12-Tricosadiynoic Acid
CAS 66990-30-5
Molekylær formel C23H38O2
Mængde 1 mg
InChI formel InChI=1S/C23H38O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25/h2-10,15-22H2,1H3,(H,24,25)
SMIL CCCCCCCCCCC#CC#CCCCCCCCCC(=O)O
IUPAC navn Trikosa-10,12-diynsyre
Molekylvægt (g/mol) 346.56
Formel vægt 346.2872
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.