Få mere at vide
10,11-Dihydro-5H-dibenzo[a,d]cycloheptene (Dibenzosuberane), Mikromol™
Artikelnummer. 30051560
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051560 25 mg
30049935 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051560 Leverandør Mikromol Leverandørnr. MM0062.130025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
10,11-Dihydro-5H-dibenzo[a,d]cycloheptene (Dibenzosuberane)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 10,11-Dihydro-5H-dibenzo[a,d]cycloheptene (Dibenzosuberane)
CAS 833-48-7
Analyt- eller komponentnavne 10,11-Dihydro-5H-dibenzo[a,d]cycloheptene (Dibenzosuberane)
Forsendelsestilstand Room Temperature
Molekylær formel C15 H14
Certificeringer/compliance ISO 17025
Synonym 10,11-Dihydro-5H-dibenzo[a,d]cycloheptene, Dibenzosuberane
InChI formel InChI=1S/C15H14/c1-3-7-14-11-15-8-4-2-6-13(15)10-9-12(14)5-1/h1-8H,9-11H2
SMIL C1Cc2ccccc2Cc3ccccc13
IUPAC navn 6,11-dihydro-5H-dibenzo[2,1-b:1',2'-e][7]annulene
Molekylvægt (g/mol) 194.27
Mængde 25 mg
Formel vægt 194.1096 g/mol