Få mere at vide
10,11-Dihydro-10-hydroxy Carbamazepine-d3, TRC
Artikelnummer. 30378176
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30378176 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378176 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4491381MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 10,11-Dihydro-10-hydroxycarbazepine-d3
CAS 1189917-36-9
Renhedsgrad noter HPLC
Molekylær formel C15 2H3 H11 N2 O2
Mængde 1 mg
Synonym (R)-10-Monohydroxy-10,11-Dihydro Carbamazepine-D3
InChI formel InChI=1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19)/i9D2,14D
SMIL [2H]C1([2H])c2ccccc2N(C(=O)N)c3ccccc3C1([2H])O
IUPAC navn 5,6,6-trideuterio-5-hydroxybenzo[b][1]benzazepine-11-carboxamide
Molekylvægt (g/mol) 257.3
Formel vægt 257.1244
Procent renhed >95