Få mere at vide
1-Tetratriacontanol-d4, TRC
Artikelnummer. 30455269
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30455269 2.5 mg 2.5mg
30454158 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30455269 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3064822.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Tetratriacontanol-d4
CAS 1346602-70-7
Molekylær formel C34D4H66O
Mængde 2.5 mg
Synonym Geddyl Alcohol-d4, Sapiol-d4, Tetratriacontyl Alcohol-d4
InChI formel InChI=1S/C34H70O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30-31-32-33-34-35/h35H,2-34H2,1H3/i31D2,32D2
SMIL OCCC([2H])([2H])C([2H])([2H])CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Molekylvægt (g/mol) 498.94
Formel vægt 498.5678