missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Tetradecyne, tech. 90%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1-Tetradecyne, is used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Tetradecyne, is used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.
Solubility
Not miscible with water. soluble in common organic solvents of low polarity
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
1-Tetradecyne, is used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.
Solubility
Not miscible with water. soluble in common organic solvents of low polarity
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Specifications
Density | 0.79 |
Boiling Point | 132°C to 134°C (14 mmHg) |
Refractive Index | 1.441 |
Quantity | 5 g |
Beilstein | 1746546 |
Solubility Information | Not miscible with water. soluble in common organic solvents of low polarity |
Formula Weight | 194.36 |
Percent Purity | 90% |
Grade | Technical |
Chemical Name or Material | 1-Tetradecyne |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'