Få mere at vide
1-(tert-Butyldimethylsilyloxy)-1-ethynyl-cyclopropane-d4, TRC
Artikelnummer. 30334618
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30334618 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334618 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6914572.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(tert-Butyldimethylsilyloxy)-1-ethynyl-cyclopropane-d4
CAS 1246817-59-3
Molekylær formel C11H16D4OSi
Mængde 2.5 mg
Synonym 1-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-1-ethynyl-cyclopropane-d4, (1,1-Dimethylethyl)[(1-ethynylcyclopropyl)oxy]dimethyl-silane-d4
InChI formel InChI=1S/C11H20OSi/c1-7-11(8-9-11)12-13(5,6)10(2,3)4/h1H,8-9H2,2-6H3/i8D2,9D2
SMIL [2H]C1([2H])C([2H])([2H])C1(O[Si](C)(C)C(C)(C)C)C#C
IUPAC navn tert-butyl-dimethyl-(2,2,3,3-tetradeuterio-1-ethynylcyclopropyl)oxysilane
Molekylvægt (g/mol) 200.39
Formel vægt 200.1534