Learn More
1-(tert-butyl) 6-Methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1,6-dicarboxylate, 98%, BLD Pharm™
Product Code. 30948348
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
Unit Size:
100mg
1g
250mg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948348 250 mg 250mg
30935076 25 g 25g
30945777 100 mg 100mg
30932043 1 g 1g
30943381 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30948348 Supplier BLD Pharm Supplier No. BD00794072250mg
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 2377609-81-7
Molecular Formula C21H28BNO6
MDL Number MFCD30717679
Quantity 250 mg
SMILES O=C(C1=CC2=C(C=C1)C(B3OC(C)(C)C(C)(C)O3)=CN2C(OC(C)(C)C)=O)OC
Molecular Weight (g/mol) 401.26
Percent Purity 98%
Chemical Name or Material 1-(tert-butyl) 6-Methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1,6-dicarboxylate