Få mere at vide
1-Stearoyl-sn-glycerol, TRC
Artikelnummer. 30413699
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30413699 2.5 g 2.5g
30410058 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30413699 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6865652.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22610-61-3
Molekylær formel C21H42O4
Molekylvægt (g/mol) 358.56
Synonym Octadecanoic Acid (2S)-2,3-Dihydroxypropyl Ester, L-(+)-1-Monostearin, 1-O-Octadecanoyl-2n-glycerol
IUPAC navn [(2S)-2,3-dihydroxypropyl] octadecanoate
SMIL CCCCCCCCCCCCCCCCCC(=O)OC[C@H](CO)O
Tekniske data
Kemisk navn eller materiale 1-Stearoyl-sn-glycerol
Mængde 2.5 g
InChI formel InChI=1S/C21H42O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h20,22-23H,2-19H2,1H3/t20-/m0/s1
Formel vægt 358.3083