Få mere at vide
1-Stearoyl-rac-glycerol-d5, TRC
Artikelnummer. 30416759
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30416759 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416759 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68656325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Stearoyl-rac-glycerol-d5
CAS 1432065-28-5
Molekylær formel C21H27D5O4
Mængde 25 mg
Synonym Octadecanoic Acid 2,3-Dihydroxypropyl Ester-d5, 1-Monostearin-d5, (+/-)-2,3-Dihydroxypropyl Octadecanoate-d5, 1-Glyceryl Stearate-d5, 1-Monooctadecanoylglycerol-d5, Aldo 33-d5, Aldo 75-d5, Aldo MSD-d5, NSC 3875-d5, Sandin EU-d5, Stearic Acid α-Monoglyceride-d5, Stearic Acid 1-Monoglyceride-d5, Tegin 55G-d5, α-Monostearin-d5
InChI formel InChI=1S/C21H42O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h20,22-23H,2-19H2,1H3/i18D2,19D2,20D
SMIL OC([2H])([2H])C(O)([2H])C([2H])([2H])OC(CCCCCCCCCCCCCCCCC)=O
Molekylvægt (g/mol) 363.59
Formel vægt 363.3397