missing translation for 'onlineSavingsMsg'
Få mere at vide

(1’S,2'S)-Nicotine 1'-Oxide and (1’R,2'S)-Nicotine 1'-Oxide Mixture, TRC

Product Code. 18010450
Change view
Click to view available options
Mængde:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18010450 10 mg
18022650 250 mg
18042350 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18010450 Supplier Toronto Research Chemicals Supplier No. TRCN42751010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-26-9
Molekylær formel C10 H14 N2 O
Molekylvægt (g/mol) 178.231
Synonym Pyridine, 3-[(2S)-1-methyl-1-oxido-2-pyrrolidinyl]-, Nicotine, 1'-oxide (7CI,8CI), Pyridine, 3-(1-methyl-2-pyrrolidinyl)-, N-oxide, (2S)-, 3-[(2S)-1-Methyl-1-oxido-2-pyrrolidinyl]pyridine, (-)-Nicotine 1'-N-oxide, (-)-Nicotine 1'-oxide, (2S)-1-Methyl-2-(pyridin-3-yl)pyrrolidin-1-ium-1-olate, N-Oxynicotine, Nicotine 1'-N-oxide, Nicotine-N'-oxide, Oxynicotine, Pyridine, 3-(1-methyl-1-oxido-2-pyrrolidinyl)-, (2S)-, S-(-)-Nicotine-1'-N-oxide, S-Nicotine-N-oxide, (1RS,2S)-1-Methyl-2-(pyridin-3-yl)pyrrolidine 1-oxide, Nicotine N'-oxide
IUPAC navn 3-[(2S)-1-methyl-1-oxidopyrrolidin-1-ium-2-yl]pyridine
SMIL C[N+]1([O-])CCC[C@H]1c2cccnc2

Specifications

Kemisk navn eller materiale (1RS,2S)-1-Methyl-2-(pyridin-3-yl)pyrrolidine 1-Oxide (Nicotine N'-Oxide)
Renhedsgrad noter GC
Mængde 10 mg
InChI formel InChI=1S/C10H14N2O/c1-12(13)7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/t10-,12?/m0/s1
Formel vægt 178.1106
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.