missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrimidin-2-yl-piperidine-4-carboxylic acid, Thermo Scientific™

Catalog Number p-4391465
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
303144-44-7
C10H12N3O2
206.23
MFCD00664474
DYVXURZASBPYFR-UHFFFAOYSA-M
1-pyrimidin-2-yl piperidine-4-carboxylic acid, 1-pyrimidin-2-yl-piperidine-4-carboxylic acid, 1-2-pyrimidinyl piperidine-4-carboxylic acid, 1-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-2-pyrimidyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-2-pyrimidinyl, acmc-1agno, 4-piperidinecarboxylicacid, 1-2-pyrimidinyl
2767267
1-pyrimidin-2-ylpiperidine-4-carboxylic acid
[O-]C(=O)C1CCN(CC1)C1=NC=CC=N1
This item is not returnable. View return policy
303144-44-7
C10H12N3O2
206.23
MFCD00664474
DYVXURZASBPYFR-UHFFFAOYSA-M
1-pyrimidin-2-yl piperidine-4-carboxylic acid, 1-pyrimidin-2-yl-piperidine-4-carboxylic acid, 1-2-pyrimidinyl piperidine-4-carboxylic acid, 1-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-2-pyrimidyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-2-pyrimidinyl, acmc-1agno, 4-piperidinecarboxylicacid, 1-2-pyrimidinyl
2767267
1-pyrimidin-2-ylpiperidine-4-carboxylic acid
[O-]C(=O)C1CCN(CC1)C1=NC=CC=N1
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 207.23
Percent Purity 97%
Chemical Name or Material 1-Pyrimidin-2-yl-piperidine-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'