Få mere at vide
1-Pyrenol-d9 Acetate, TRC
Artikelnummer. 30421213
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30421213 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421213 Leverandør Toronto Research Chemicals Leverandørnr. TRCP98958210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Pyrenol-d9 Acetate
CAS 1246820-93-8
Molekylær formel C18 H3 D9 O2
Mængde 10 mg
Synonym 1-Acetoxypyrene-d9
InChI formel InChI=1S/C18H12O2/c1-11(19)20-16-10-8-14-6-5-12-3-2-4-13-7-9-15(16)18(14)17(12)13/h2-10H,1H3/i2D,3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c2c([2H])c([2H])c3c([2H])c([2H])c(OC(=O)C)c4c([2H])c([2H])c(c1[2H])c2c34
IUPAC navn (2,3,4,5,6,7,8,9,10-nonadeuteriopyren-1-yl) acetate
Molekylvægt (g/mol) 269.34
Formel vægt 269.1402