missing translation for 'onlineSavingsMsg'
Learn More

1-(Pyren-1-yl)-1H-pyrrole-2,5-dione, 98%, BLD Pharm™

Product Code. 30981529
Encompass_Preferred
Change view
Click to view available options
Mængde:
100 mg
250 mg
1 g
5 g
Unit Size:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde Unit Size
30981529 5 g -
30977148 100 mg 100mg
30979475 250 mg -
30976081 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30981529 Supplier BLD Pharm Supplier No. BD1407145g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale 1-(Pyren-1-yl)-1H-pyrrole-2,5-dione
CAS 42189-56-0
Molekylær formel C20H11NO2
MDL nummer MFCD00049301
Mængde 5 g
SMIL O=C(N1C2=C(C3=C45)C=CC5=CC=CC4=CC=C3C=C2)C=CC1=O
Molekylvægt (g/mol) 297.31
Procent renhed 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.