Få mere at vide
1-Propylamine-d3, TRC
Artikelnummer. 30427280
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30427280 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427280 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8337031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Propylamine-d3
CAS 24300-22-9
Molekylær formel C3 D3 H6 N
Mængde 1 mg
Synonym 1-Propan-3,3,3-d3-amine (9CI), Propyl-3,3,3-d3-amine (7CI','8CI), n-Propyl-3,3,3-d3-amine, 1-Propylamine-d3, Propylamine-D3 (propyl-3,3,3-D3)
InChI formel InChI=1S/C3H9N/c1-2-3-4/h2-4H2,1H3/i1D3
SMIL [2H]C([2H])([2H])CCN
IUPAC navn 3,3,3-trideuteriopropan-1-amine
Molekylvægt (g/mol) 62.129
Formel vægt 62.092