Få mere at vide
1-Propanethiol-SD, CDN
Artikelnummer. 30319242
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30319242 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319242 Leverandør CDN Leverandørnr. CDND02221G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Propanethiol-S-D
CAS 64071-73-4
Molekylær formel C3 D H7 S
Mængde 1 g
Synonym 1-Propanethiol-d (9 CI), 1-Propanethiol-SD, 1-Propanethiol-S-D, 1-Mercapto-d-propane, 1-Propyl mercaptan-d
InChI formel InChI=1 S/C3H8S/c1-2-3-4/h4H,2-3H2,1H3/i/hD
SMIL [2 H]SCCC
IUPAC navn 1-deuteriosulfanylpropane
Molekylvægt (g/mol) 77.1668
Formel vægt 77.0409 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity