Få mere at vide
1-Phenylpentadecane-d36, CDN
Artikelnummer. 30322074
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322074 0.1 g 0.1g
30319596 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322074 Leverandør CDN Leverandørnr. CDND52310.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Phenylpentadecane-D36
CAS 352431-31-3
Molekylær formel C21 D36
Mængde 0.1 g
Synonym Benzene-d5, pentadecyl-d31- (9 CI), 1-Phenylpentadecane-d36, 1-Phenylpentadecane-D36, Pentadecyl-d31-benzene-d5, Perdeuteropentadecylbenzene
InChI formel InChI=1 S/C21H36/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-18-21-19-16-14-17-20-21/h14,16-17,19-20 H,2-13,15,18H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14 D,15D2,16 D,17 D,18D2,19 D,20 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,15-hentriacontadeuteriopentadecyl)benzene
Molekylvægt (g/mol) 324.7324
Formel vægt 324.5077 g/mol
Procent renhed 98 atom % D, min 97% Chemical Purity