Få mere at vide
(±)-1-Phenyl-d5-ethanol, TRC
Artikelnummer. 30473560
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30473560 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473560 Leverandør Toronto Research Chemicals Leverandørnr. TRCM226187100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Phenylethanol-D5 (phenyl-D5)
CAS 90162-45-1
Molekylær formel C8 D5 H5 O
Mængde 100 mg
Synonym Benzene-2,3,4,5,6-d5-methanol, α-methyl- (ACI), Benzene-d5-methanol, α-methyl- (9CI), α-Methylbenzene-2,3,4,5,6-d5-methanol (ACI), 1-(2,3,4,5,6-Pentadeuteriophenyl)ethanol, 1-(Phenyl-d5)ethanol, 1-Phenylethanol-D5 (phenyl-D5), 1-Phenylethanol-d5, (±)-1-Phenyl-d5-ethanol
InChI formel InChI=1S/C8H10O/c1-7(9)8-5-3-2-4-6-8/h2-7,9H,1H3/i2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(C(C)O)c([2H])c1[2H]
IUPAC navn 1-(2,3,4,5,6-pentadeuteriophenyl)ethanol
Molekylvægt (g/mol) 127.1952
Formel vægt 127.1045