missing translation for 'onlineSavingsMsg'
Få mere at vide

1-Phenyl-2-(4-N-Cbz-phenylamino-piperidin-1-yl)-propan-1-one, TRC

Artikelnummer. 30411655
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30411655 100 mg
30428638 50 mg
30418859 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30411655 Supplier Toronto Research Chemicals Supplier No. TRCP336948100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 936498-12-3
Molekylær formel C28H30N2O3
Molekylvægt (g/mol) 442.56
IUPAC navn benzyl N-[1-(1-oxo-1-phenylpropan-2-yl)piperidin-4-yl]-N-phenylcarbamate
SMIL CC(N1CCC(CC1)N(C(=O)OCc2ccccc2)c3ccccc3)C(=O)c4ccccc4

Specifications

Kemisk navn eller materiale 1-Phenyl-2-(4-N-Cbz-phenylamino-piperidin-1-yl)-propan-1-one
Mængde 100 mg
InChI formel InChI=1S/C28H30N2O3/c1-22(27(31)24-13-7-3-8-14-24)29-19-17-26(18-20-29)30(25-15-9-4-10-16-25)28(32)33-21-23-11-5-2-6-12-23/h2-16,22,26H,17-21H2,1H3
Formel vægt 442.2256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.