missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Phenyl-1H-1,2,3-triazole-4-methanol, 97%

Brand:  Alfa Aesar™ H64892.MD

Additional Details : CAS Number : 103755-58-4 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15402299

  • 229.22 DKK / 250mg


Estimated Shipment date
from Supplier 07-05-2018
Add to basket

Specifications

Chemical Identifiers

1-Phenyl-1H-1,2,3-triazole-4-methanol
175.19
145841
103755-58-4
MFCD00100214
4-Hydroxymethyl-1-phenyl-1H-1,2,3-triazole

Specifications

1-Phenyl-1H-1,2,3-triazole-4-methanol
103755-58-4
97%
MFCD00100214
4-Hydroxymethyl-1-phenyl-1H-1,2,3-triazole
250mg
Solid
175.19
145841
Videos
SDS