Få mere at vide
1-Phenyl-1-butyne-3,3,4,4,4-d5, CDN
Artikelnummer. 30310177
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30310177 0.5 g
30317060 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310177 Leverandør CDN Leverandørnr. CDND68060.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Phenyl-1-butyne-3,3,4,4,4-D5
CAS 1219803-38-9
Molekylær formel C10 D5 H5
Mængde 0.5 g
Synonym Benzene, 1-butyn-1-yl-3,3,4,4,4-d5- (ACI), 1-Butyn-1-yl-3,3,4,4,4-d5benzene (ACI), 1-Phenyl-1-butyne-3,3,4,4,4-d5, 1-Phenyl-1-butyne-3,3,4,4,4-D5, 1-(Ethyl-d5)-2-phenylacetylene
InChI formel InChI=1 S/C10H10/c1-2-3-7-10-8-5-4-6-9-10/h4-6,8-9 H,2H2,1H3/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C#Cc1ccccc1
IUPAC navn 3,3,4,4,4-pentadeuteriobut-1-ynylbenzene
Molekylvægt (g/mol) 135.2172
Formel vægt 135.1096 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity