Få mere at vide
1-Phenanthrol-d9, TRC
Artikelnummer. 30410465
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30410465 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410465 Leverandør Toronto Research Chemicals Leverandørnr. TRCP29500210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Phenanthrol-d9
CAS 922510-23-4
Renhedsgrad noter HPLC
Molekylær formel C14HD9O
Mængde 10 mg
Synonym Phenanthren-2,3,4,5,6,7,8,9,10-d9-1-ol, 1-Phenanthrenol-d9, 1-Hydroxyphenanthrene-d9, NSC 44471-d9
InChI formel InChI=1S/C14H10O/c15-14-7-3-6-12-11-5-2-1-4-10(11)8-9-13(12)14/h1-9,15H/i1D,2D,3D,4D,5D,6D,7D,8D,9D
SMIL OC1=C([2H])C([2H])=C([2H])C2=C1C([2H])=C([2H])C3=C([2H])C([2H])=C([2H])C([2H])=C32
IUPAC navn 2,3,4,5,6,7,8,9,10-nonadeuteriophenanthren-1-ol
Molekylvægt (g/mol) 203.28
Formel vægt 203.1297
Procent renhed >95