Få mere at vide
1-Oxyl-2,2,5,5-tetramethyl-∆3-(methanesulfonyloxymethyl)pyrroline, TRC
Artikelnummer. 30427147
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427147 25 mg
30428318 10 mg
30416540 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30427147 Leverandør Toronto Research Chemicals Leverandørnr. TRCO87240025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76893-27-1
Molekylær formel C10 H18 N O4 S
Molekylvægt (g/mol) 248.32
IUPAC navn (1-λ^{1}-oxidanyl-2,2,5,5-tetramethylpyrrol-3-yl)methyl methanesulfonate
SMIL CC1(C)C=C(COS(=O)(=O)C)C(C)(C)N1[O]
Tekniske data
Kemisk navn eller materiale 1-Oxyl-2,2,5,5-tetramethyl-delta3-(methanesulfonyloxymethyl)pyrroline
Mængde 25 mg
InChI formel InChI=1S/C10H18NO4S/c1-9(2)6-8(7-15-16(5,13)14)10(3,4)11(9)12/h6H,7H2,1-5H3
Formel vægt 248.0957