Få mere at vide
1-Octanol-d17, TRC
Artikelnummer. 30428400
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30428400 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30428400 Leverandør Toronto Research Chemicals Leverandørnr. TRCO237902500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Octanol-d17
CAS 153336-13-1
Molekylær formel C8 D17 H O
Mængde 500 mg
Synonym 1-Octan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d17-ol, 1-Octan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d17-ol (9CI), 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecadeuteriooctan-1-ol, Octyl Alcohol-d17, 1-Hydroxyoctane-d17, Alfol 8-d17, CO 898-d17, CO 898 (solvent)-d17, Caprylic Alcohol-d17, Heptyl Carbinol-d17, Kalcohl 0898-d17, Kalcol 0898-d17, Lorol C 8-98-d17, NSC 9823-d17, Octanol-d17, Octilin-d17, Sipol L8-d17, n-Octan-1-ol-d17, n-Octanol-d17, n-Octyl Alcohol-d17, n-Octyl-d17 alcohol
InChI formel InChI=1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])O
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecadeuteriooctan-1-ol
Molekylvægt (g/mol) 147.33
Formel vægt 147.2425