missing translation for 'onlineSavingsMsg'
Få mere at vide

1-O-n-Octyl-d17-beta-D-glucopyranoside, CDN

Artikelnummer. 30308773
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.01 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30308773 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308773 Leverandør CDN Leverandørnr. CDND52600.01G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Octyl beta-D-Glucopyranoside-D17 (octyl-D17)
CAS 129522-81-2
Molekylær formel C14 D17 H11 O6
Mængde 0.01 g
Synonym β-D-Glucopyranoside, octyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d17 (ACI), Octyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d17 β-D-glucopyranoside (ACI), β-D-Glucopyranoside, octyl-d17 (9 CI), Octyl-d17 β-D-glucopyranoside, Octyl β-D-glucopyranoside-D17 (octyl-D17), 1-O-(Octyl-d17)-β-D-glucopyranoside
InChI formel InChI=1 S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18 H,2-9H2,1H3/t10-,11-,12+,13-,14-/m1/s1/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O[C@@H]1 O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1 O
IUPAC navn (2 R,3 R,4 S,5 S,6 R)-2-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecadeuteriooctoxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Molekylvægt (g/mol) 309.4733
Formel vægt 309.2953 g/mol
Procent renhed 98 atom % D, min 97% Chemical Purity
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.