Få mere at vide
1-O-Alpha-D-Glucopyranosyl-D-mannitol, TRC
Artikelnummer. 30315333
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315333 250 mg
30319431 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30315333 Leverandør Toronto Research Chemicals Leverandørnr. TRCA576035250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-O-alpha-D-Glucopyranosyl-D-mannitol
CAS 20942-99-8
Molekylær formel C12H24O11
Mængde 250 mg
Synonym 6-O-alpha-D-Glucopyranosyl-D-mannitol, 1,1-GPM
InChI formel InChI=1S/C12H24O11/c13-1-4(15)7(17)8(18)5(16)3-22-12-11(21)10(20)9(19)6(2-14)23-12/h4-21H,1-3H2/t4-,5-,6-,7-,8-,9-,10+,11-,12+/m1/s1
SMIL OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
IUPAC navn (2R,3R,4R,5R)-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,4,5-pentol
Molekylvægt (g/mol) 344.31
Formel vægt 344.1319