Få mere at vide
1-Nonadecanol, Mikromol™
Artikelnummer. 30052433
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052433 100 mg
30049178 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052433 Leverandør Mikromol Leverandørnr. MM3450.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Nonadecanol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Nonadecanol
CAS 1454-84-8
Analyt- eller komponentnavne 1-Nonadecanol
Forsendelsestilstand Room Temperature
Molekylær formel C19 H40 O
Certificeringer/compliance ISO 17025
Synonym 1-Nonadecanol, Nonadecyl alcohol, n-1-Nonadecanol, n-Nonadecanol
InChI formel InChI=1S/C19H40O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20/h20H,2-19H2,1H3
SMIL CCCCCCCCCCCCCCCCCCCO
IUPAC navn nonadecan-1-ol
Molekylvægt (g/mol) 284.52
Mængde 100 mg
Formel vægt 284.3079 g/mol